missing translation for 'onlineSavingsMsg'
Learn More

DL-Valinol 98.0+%, TCI America™

Catalog No. V01005G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01005G 5 g
V01001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01005G Supplier TCI America Supplier No. V01005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16369-05-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00004730
InChI Key NWYYWIJOWOLJNR-UHFFFAOYSA-N
Synonym 2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol
PubChem CID 79019
IUPAC Name 2-amino-3-methylbutan-1-ol
SMILES CC(C)C(CO)N

Specifications

Color Yellow
Boiling Point 87°C
Quantity 5 g
Formula Weight 103.17
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material DL-Valinol