missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine 98.0+%, TCI America™

Catalog No. p-7137886
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V001325G 25 g
V0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V001325G Supplier TCI America Supplier No. V001325G

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYSA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O

Specifications

Melting Point 295°C
Color White
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Valine
EINECSNumber (9)-1604
TSCA Yes