missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine ethyl ester hydrochloride, 99%

Catalog No. p-7047185
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1790606 5 g
AAL1790614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1790606 Supplier Thermo Scientific Chemicals Supplier No. L1790606
Only null left

CAS: 23358-42-1 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23358-42-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00067542
InChI Key PQGVTLQEKCJXKF-UHFFFAOYSA-N
Synonym dl-valine ethyl ester hydrochloride, ethyl 2-amino-3-methylbutanoate hydrochloride, h-dl-val-oet.hcl, ethyl dl-valinate hydrochloride, valine ethyl ester, hydrochloride, ethyl dl-valinate hcl, acmc-20al3t, acmc-209ta5, +/--valine ethyl ester hydrochloride, ethyl 2-azanyl-3-methyl-butanoate hydrochloride
PubChem CID 13000942
IUPAC Name ethyl 2-amino-3-methylbutanoate;hydrochloride
SMILES Cl.CCOC(=O)C(N)C(C)C

Specifications

Melting Point ∼94°C
Beilstein 3912411
Quantity 5 g
Formula Weight 181.66
Percent Purity 99%
Chemical Name or Material DL-Valine ethyl ester hydrochloride
EINECSNumber 245-606-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only