missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Valine, 99%
CAS: 516-06-3 | C5H11NO2 | 117.15 g/mol
$80.96 - $1121.18
Chemical Identifiers
| CAS | 516-06-3 |
|---|---|
| Molecular Formula | C5H11NO2 |
| Molecular Weight (g/mol) | 117.15 |
| MDL Number | MFCD00004267 |
| InChI Key | KZSNJWFQEVHDMF-UHFFFAOYSA-N |
| Synonym | dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid |
| PubChem CID | 1182 |
| ChEBI | CHEBI:27266 |
| IUPAC Name | 2-amino-3-methylbutanoic acid |
| SMILES | CC(C)C(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1675622
|
Thermo Scientific Chemicals
A1675622 |
100 g |
Each for $80.96
|
|
|||||
|
AAA1675636
|
Thermo Scientific Chemicals
A1675636 |
500 g |
Each for $257.66
|
|
|||||
|
AAA167560E
|
Thermo Scientific Chemicals
A167560E |
2500 g |
Each for $1,121.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 516-06-3 | |
| 117.15 | |
| KZSNJWFQEVHDMF-UHFFFAOYSA-N | |
| 1182 | |
| 2-amino-3-methylbutanoic acid |
| C5H11NO2 | |
| MFCD00004267 | |
| dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid | |
| CHEBI:27266 | |
| CC(C)C(N)C(O)=O |
Specifications
| ∼296°C (sublimation) | |
| C5H11NO2 | |
| 506689 | |
| dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid | |
| CC(C)C(N)C(O)=O | |
| 100 g | |
| 1182 | |
| 117.15 | |
| DL-Valine |
| 516-06-3 | |
| MFCD00004267 | |
| 14,9909 | |
| KZSNJWFQEVHDMF-UHFFFAOYSA-N | |
| 2-amino-3-methylbutanoic acid | |
| 117.15 | |
| CHEBI:27266 | |
| 99% |
Safety and Handling
EINECSNumber : 208-220-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only