missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Valine, 99+%, for biochemistry
CAS: 516-06-3 | C5H11NO2 | 117.15 g/mol
Supplier: Thermo Scientific Chemicals 172102500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistrySpecifications
| 283.5°C to 285.0°C | |
| 99+% | |
| White | |
| 10ppm max. | |
| 0.2% max. (105°C, 3 hrs) | |
| C5H11NO2 | |
| (CH3)2CHCH(NH2)CO2H | |
| 04, 430 | |
| 14, 9909 | |
| dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid | |
| KZSNJWFQEVHDMF-UHFFFAOYSA-N | |
| CC(C)C(N)C(O)=O | |
| 117.15 | |
| 1182 | |
| 117.15 | |
| 1.31 | |
| Crystalline Powder |
| DL-Valine | |
| 516-06-3 | |
| 400ppm max. | |
| Authentic | |
| 99+% | |
| Plastic Bottle | |
| MFCD00004267 | |
| 95% min. (c=10, 2N HCl, 430nm) 1cm cell | |
| 0.2% max. | |
| Solubility in water: 68g/L. Other solubilities: practically insoluble in cold alcohol and ether, soluble in mineral acids, insoluble in organic solvents, slightly soluble in benzene, ethanol | |
| 1.3100g/mL | |
| 2-amino-3-methylbutanoic acid | |
| 250 g | |
| CHEBI:27266 | |
| ≥99% | |
| Biochemical |
Chemical Identifiers
| 516-06-3 | |
| 117.15 | |
| KZSNJWFQEVHDMF-UHFFFAOYSA-N | |
| 1182 | |
| 2-amino-3-methylbutanoic acid |
| C5H11NO2 | |
| MFCD00004267 | |
| dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid | |
| CHEBI:27266 | |
| CC(C)C(N)C(O)=O |
Safety and Handling
EINECSNumber : 208-220-0
TSCA : TSCA
RUO – Research Use Only