missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Valine, 99+%, for biochemistry

Catalog No. AC172100500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172100500 50 g Plastic Bottle
AC172102500 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172100500 Supplier Thermo Scientific Chemicals Supplier No. 172100500

CAS: 516-06-3 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYSA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O

Specifications

Melting Point 283.5°C to 285.0°C
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Loss on Drying 0.2% max. (105°C, 3 hrs)
Assay Percent Range 99+%
Packaging Plastic Bottle
Linear Formula (CH3)2CHCH(NH2)CO2H
Beilstein 04, 430
Merck Index 14, 9909
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: 68g/L. Other solubilities: practically insoluble in cold alcohol and ether, soluble in mineral acids, insoluble in organic solvents, slightly soluble in benzene, ethanol
Density 1.3100g/mL
Quantity 50 g
Formula Weight 117.15
Percent Purity ≥99%
Grade Biochemical
Specific Gravity 1.31
Physical Form Crystalline Powder
Chemical Name or Material DL-Valine, 99+%
Show More Show Less
EINECSNumber 208-220-0
TSCA TSCA

RUO – Research Use Only