missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Tropic Acid 98.0+%, TCI America™
Supplier: TCI America T0533250G
Specifications
| DL-Tropic Acid | |
| 118°C | |
| C9H10O3 | |
| 250 g | |
| JACRWUWPXAESPB-UHFFFAOYNA-N | |
| 3-hydroxy-2-phenylpropanoic acid | |
| 10726 | |
| 166.18 | |
| Crystalline Powder |
| 552-63-6 | |
| White | |
| MFCD00004255 | |
| tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
| OCC(C(O)=O)C1=CC=CC=C1 | |
| 166.18 | |
| CHEBI:30765 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 552-63-6 | |
| 166.18 | |
| JACRWUWPXAESPB-UHFFFAOYNA-N | |
| 10726 | |
| 3-hydroxy-2-phenylpropanoic acid |
| C9H10O3 | |
| MFCD00004255 | |
| tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
| CHEBI:30765 | |
| OCC(C(O)=O)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-3083
RTECSNumber : CY8535000
TSCA : Yes