missing translation for 'onlineSavingsMsg'
Learn More

DL-Serine, Non-Animal, ≥98.5%

Catalog No. p-7082826
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ2148622 100 g Plastic Bottle
AAJ21486A1 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ2148622 Supplier Thermo Fisher Scientific Supplier No. J2148622
Only null left

CAS: 302-84-1 | C3H7NO3 | 105.093 g/mol

Chemical Identifiers

CAS 302-84-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064223
InChI Key MTCFGRXMJLQNBG-UHFFFAOYSA-N
Synonym dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid
PubChem CID 617
ChEBI CHEBI:17822
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point ∼240°C (decomposition)
Packaging Plastic Bottle
Linear Formula CH2OHCHNH2COOH
Beilstein 1721402
Quantity 100 g
Formula Weight 105.09
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material DL-Serine
EINECSNumber 206-130-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only