missing translation for 'onlineSavingsMsg'
Learn More

DL-Serine methyl ester hydrochloride, 98+%

Catalog No. AAA1390430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1390430 250 g
AAA1390409 10 g
AAA1390418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1390430 Supplier Thermo Scientific Chemicals Supplier No. A1390430

CAS: 5619-04-5 | C4H10ClNO3 | 155.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5619-04-5
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.58
MDL Number MFCD00012593
InChI Key NDBQJIBNNUJNHA-UHFFFAOYNA-N
Synonym dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908
PubChem CID 517373
IUPAC Name methyl 2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point ∼134°C (decomposition)
Beilstein 6067970
Quantity 250 g
Formula Weight 155.58
Percent Purity ≥98%
Sensitivity Moisture sensitive
Chemical Name or Material DL-Serine methyl ester hydrochloride
EINECSNumber 227-047-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only