missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Serine methyl ester hydrochloride, 98+%
CAS: 5619-04-5 | C4H10ClNO3 | 155.58 g/mol
Supplier: Thermo Scientific Chemicals A1390418
| Quantity | 50 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C4H10ClNO3 | |
| MFCD00012593 | |
| dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908 | |
| methyl 2-amino-3-hydroxypropanoate;hydrochloride |
Specifications
| ∼134°C (decomposition) | |
| 5619-04-5 | |
| MFCD00012593 | |
| dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908 | |
| COC(=O)C(CO)N.Cl | |
| 50 g | |
| 517373 | |
| ≥98% |
| DL-Serine methyl ester hydrochloride | |
| C4H10ClNO3 | |
| 6067970 | |
| NDBQJIBNNUJNHA-UHFFFAOYNA-N | |
| methyl 2-amino-3-hydroxypropanoate;hydrochloride | |
| 155.58 | |
| 155.58 | |
| Moisture sensitive |
Safety and Handling
EINECSNumber : 227-047-1
TSCA : No
Recommended Storage : Keep cold