Learn More
DL-sec-Phenethyl alcohol, 97%
CAS: 98-85-1 | C8H10O | 122.17 g/mol
$64.10 - $168.75
Chemical Identifiers
| CAS | 98-85-1 |
|---|---|
| Molecular Formula | C8H10O |
| Molecular Weight (g/mol) | 122.17 |
| MDL Number | MFCD00004508 |
| InChI Key | WAPNOHKVXSQRPX-UHFFFAOYSA-N |
| Synonym | methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene |
| PubChem CID | 7409 |
| ChEBI | CHEBI:669 |
| IUPAC Name | 1-phenylethanol |
| SMILES | CC(C1=CC=CC=C1)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC351881000
|
Thermo Scientific Chemicals
351881000 |
100 g | Glass bottle |
Each for $64.10
|
|
||||
|
AC351885000
|
Thermo Scientific Chemicals
351885000 |
500 g | Glass bottle |
Each for $168.75
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 98-85-1 | |
| 122.17 | |
| WAPNOHKVXSQRPX-UHFFFAOYSA-N | |
| 7409 | |
| 1-phenylethanol |
| C8H10O | |
| MFCD00004508 | |
| methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene | |
| CHEBI:669 | |
| CC(C1=CC=CC=C1)O |
Specifications
| 98-85-1 | |
| Colorless | |
| 203°C to 205°C | |
| Authentic | |
| Glass bottle | |
| 1.5260 to 1.528 | |
| MFCD00004508 | |
| 06, II, 444 | |
| methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene | |
| WAPNOHKVXSQRPX-UHFFFAOYSA-N | |
| 1-phenylethanol | |
| 7409 | |
| CHEBI:669 | |
| 97% | |
| DL-sec-Phenethyl alcohol, 97% |
| 20°C | |
| 1.0100g/mL | |
| 86°C | |
| 96% min. (GC) | |
| C8H10O | |
| C6H5CH(CH3)OH | |
| 100 g | |
| 1.01 | |
| Solubility in water: 29g/L (20°C). Other solubilities: soluble in alcohol and glycerol | |
| CC(C1=CC=CC=C1)O | |
| 122.17 | |
| 11.2 mPa.s (20°C) | |
| 122.17 | |
| Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 202-707-1
RTECSNumber : DO9275000
TSCA : TSCA
RUO – Research Use Only