missing translation for 'onlineSavingsMsg'
Learn More

DL-Prolinol 95.0+%, TCI America™

Catalog No. P17935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17935G 5 g
P179325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17935G Supplier TCI America Supplier No. P17935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 498-63-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00601073
InChI Key HVVNJUAVDAZWCB-UHFFFAOYSA-N
Synonym 2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol #
PubChem CID 90908
ChEBI CHEBI:59652
IUPAC Name pyrrolidin-2-ylmethanol
SMILES C1CC(NC1)CO

Specifications

Color Yellow
Boiling Point 76°C
Quantity 5 g
Formula Weight 101.15
Percent Purity ≥95.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material DL-Prolinol