missing translation for 'onlineSavingsMsg'
Learn More

DL-Prolinol, 98%

Catalog No. AAH6232206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6232206 5 g
AAH6232214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6232206 Supplier Thermo Scientific Chemicals Supplier No. H6232206

CAS: 498-63-5 | C5H11NO | 101.149 g/mol

DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 498-63-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00601073
InChI Key HVVNJUAVDAZWCB-UHFFFAOYSA-N
Synonym 2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol #
PubChem CID 90908
ChEBI CHEBI:59652
IUPAC Name pyrrolidin-2-ylmethanol
SMILES C1CC(NC1)CO

Specifications

Refractive Index 1.484
Boiling Point 76°C (2mmHg)
Quantity 5 g
Solubility Information Soluble in ethanol.
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material DL-Prolinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only