missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Pipecolinic acid, 99%
Description
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.
Solubility
Soluble in water (50 mg/ml), and boiling alcohol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
DL-Pipecolinic acid is used as a diagnostic marker of pyridoxine-dependent epilepsy. It acts as a ligand in copper-catalyzed approach for P-arylation of organophosphorus compounds. It is used as organocatalyst and novel substrate in organic synthesis.
Solubility
Soluble in water (50 mg/ml), and boiling alcohol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
Specifications
Specifications
| Melting Point | ∼280°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 81095 |
| Merck Index | 14,7458 |
| Solubility Information | Soluble in water (50mg/ml),and boiling alcohol. |
| Formula Weight | 129.16 |
| Percent Purity | 99% |
| Chemical Name or Material | DL-Pipecolinic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 208-616-3 |
| RTECSNumber | TK6021000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only