missing translation for 'onlineSavingsMsg'
Learn More

DL-Pipecolic Acid 98.0+%, TCI America™

Catalog No. P044225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P044225G Supplier TCI America Supplier No. P044225G
Only null left

Chemical Identifiers

CAS 535-75-1
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00064347,MFCD00005981
InChI Key HXEACLLIILLPRG-UHFFFAOYNA-N
Synonym dl-pipecolinic acid, pipecolic acid, pipecolinic acid, 2-piperidinecarboxylic acid, homoproline, pipecolate, dl-pipecolic acid, dihydrobaikiane, piperolinic acid, hexahydropicolinic acid
PubChem CID 849
ChEBI CHEBI:17964
IUPAC Name piperidine-2-carboxylic acid
SMILES OC(=O)C1CCCCN1

Specifications

Color White
Quantity 25 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Pipecolic Acid
RTECSNumber TK6021000
TSCA Yes