missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylsuccinic acid, 98+%

Catalog No. p-3758686
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130940250 25 g Glass bottle
AC130941000 100 g Glass bottle
AC130945000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130940250 Supplier Thermo Scientific Chemicals Supplier No. 130940250
Only null left

CAS: 635-51-8 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 635-51-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004256
InChI Key LVFFZQQWIZURIO-UHFFFAOYSA-N
Synonym phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl
PubChem CID 95459
IUPAC Name 2-phenylbutanedioic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)C(=O)O

Specifications

Melting Point 166°C to 169°C
Color Cream-White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula HO2CCH2CH(C6H5)CO2H
Quantity 25 g
Beilstein 09, 865
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aceton and alcohols
Formula Weight 194.19
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material DL-Phenylsuccinic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-238-1

RUO – Research Use Only