missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylglycinol, 95%

CAS: 7568-92-5 | C8H11NO | 137.18 g/mol

$63.71 - $199.48

Chemical Identifiers

CAS 7568-92-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00130145
InChI Key IJXJGQCXFSSHNL-UHFFFAOYNA-N
Synonym 2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
PubChem CID 92466
IUPAC Name 2-amino-2-phenylethanol
SMILES NC(CO)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6393206
View Documents
Thermo Scientific Chemicals
H6393206
5 g
Each for $63.71
Only null left
 
AAH6393214
View Documents
Thermo Scientific Chemicals
H6393214
25 g
Each for $199.48
Only null left
 
Description

Description

A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.

Solubility
Soluble in chloroform, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

7568-92-5
137.18
IJXJGQCXFSSHNL-UHFFFAOYNA-N
92466
NC(CO)C1=CC=CC=C1
C8H11NO
MFCD00130145
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
2-amino-2-phenylethanol

Specifications

49°C to 53°C
White
MFCD00130145
2-phenylglycinol, dl-2-phenylglycinol, dl-phenylglycinol, 2-amino-2-phenylethan-1-ol, 2-phenyl-dl-glycinol, beta-aminophenethyl alcohol, 2-amino-2-phenyl-ethanol, benzeneethanol, beta-amino, r-2-phenylglycinol, phenethyl alcohol, beta-amino
IJXJGQCXFSSHNL-UHFFFAOYNA-N
2-amino-2-phenylethanol
5 g
137.18
Solid
7568-92-5
C8H11NO
971219
Soluble in chloroform,methanol.
NC(CO)C1=CC=CC=C1
137.18
92466
95%
DL-Phenylglycinol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

RTECSNumber : SG7600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only