missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalaninol 98.0+%, TCI America™

Catalog No. p17911g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Catalog No. Quantity
P17911G 1 g
1 options

Catalog No. P17911G

Supplier: TCI America P17911G

Only null left

Chemical Identifiers

CAS 16088-07-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00066689
InChI Key STVVMTBJNDTZBF-UHFFFAOYSA-N
Synonym dl-phenylalaninol, 2-amino-3-phenyl-1-propanol, phenylalaninol, s-phenylalaninol, l-2-amino-3-phenyl-1-propanol, s-2-benzylethanolamine, benzenepropanol, beta-amino, s-beta-aminobenzenepropanol, chembl21676, 2-amino-3-phenyl-propan-1-ol
PubChem CID 76652
IUPAC Name 2-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CC(CO)N

Specifications

Melting Point 71°C
Color White
Boiling Point 130°C
Quantity 1 g
UN Number 3259
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Phenylalaninol