missing translation for 'onlineSavingsMsg'
Learn More

DL-Pantolactone 95.0+%, TCI America™

Catalog No. p-7134374
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P001025G 25 g
P0010100G 100 g
P0010500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P001025G Supplier TCI America Supplier No. P001025G
Only null left

Chemical Identifiers

CAS 79-50-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00064333
InChI Key SERHXTVXHNVDKA-UHFFFAOYSA-N
Synonym pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
PubChem CID 989
IUPAC Name 3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 78°C
Color White
Boiling Point 121°C
Quantity 25 g
Formula Weight 130.14
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material DL-Pantolactone
EINECSNumber (9)-0997
TSCA Yes