missing translation for 'onlineSavingsMsg'
Learn More

DL-Panthenol 98.0+%, TCI America™

Catalog No. p-7135074
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P131825G 25 g
P1318500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P131825G Supplier TCI America Supplier No. P131825G
Only null left

Chemical Identifiers

CAS 16485-10-2
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00002944
InChI Key SNPLKNRPJHDVJA-UHFFFAOYNA-N
Synonym dl-panthenol, panthenol, dl-pantothenol, dl-pantothenyl alcohol, panthenolum, 2,4-dihydroxy-n-3-hydroxypropyl-3,3-dimethylbutanamide, pantenol, pantenolo, penthenol, varitan
PubChem CID 4678
IUPAC Name 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)C(O)C(=O)NCCCO

Specifications

Melting Point 66°C
Color White
Quantity 25 g
Formula Weight 205.25
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material DL-Panthenol
EINECSNumber (9)-0998
RTECSNumber ES4316500
TSCA Yes