missing translation for 'onlineSavingsMsg'
Learn More

DL-Norvaline 98.0+%, TCI America™

Catalog No. N030425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N030425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N030425G Supplier TCI America Supplier No. N030425G

Chemical Identifiers

CAS 760-78-1
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064420
InChI Key SNDPXSYFESPGGJ-UHFFFAOYSA-N
Synonym dl-norvaline, h-dl-nva-oh, dl-2-aminopentanoic acid, 2-aminovaleric acid, norvaline, dl, dl-alpha-aminovaleric acid, alpha-dl-aminopentanoic acid, norvalines, 2-aminovaleric acids, .alpha.-dl-aminopentanoic acid
PubChem CID 824
ChEBI CHEBI:19475
IUPAC Name 2-aminopentanoic acid
SMILES CCCC(N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Norvaline