missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Norvaline, 98%
Description
DL-Norvaline is a neurotransmitter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Norvaline is a neurotransmitter.
Solubility
Soluble in water.
Notes
Store in cool dry place.. Keep away from oxidizing agents.
DL-Norvaline is a neurotransmitter.
Solubility
Soluble in water.
Notes
Store in cool dry place.. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Beilstein | 1721163 |
| Merck Index | 14,6716 |
| Solubility Information | Soluble in water. |
| Quantity | 500 g |
| Formula Weight | 117.15 |
| Percent Purity | 98% |
| Chemical Name or Material | DL-Norvaline |
Safety and Handling
| EINECSNumber | 212-082-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only