missing translation for 'onlineSavingsMsg'
Learn More

DL-Norvaline, 98%

Catalog No. AAA1590022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1590022 100 g
AAA1590036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1590022 Supplier Thermo Scientific Chemicals Supplier No. A1590022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 760-78-1 | C5H11NO2 | 117.15 g/mol

DL-Norvaline is a neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Norvaline is a neurotransmitter.

Solubility
Soluble in water.

Notes
Store in cool dry place.. Keep away from oxidizing agents.

Chemical Identifiers

CAS 760-78-1
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064420
InChI Key SNDPXSYFESPGGJ-UHFFFAOYSA-N
Synonym dl-norvaline, h-dl-nva-oh, dl-2-aminopentanoic acid, 2-aminovaleric acid, norvaline, dl, dl-alpha-aminovaleric acid, alpha-dl-aminopentanoic acid, norvalines, 2-aminovaleric acids, .alpha.-dl-aminopentanoic acid
PubChem CID 824
ChEBI CHEBI:19475
IUPAC Name 2-aminopentanoic acid
SMILES CCCC(N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721163
Merck Index 14,6716
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 117.15
Percent Purity 98%
Chemical Name or Material DL-Norvaline
EINECSNumber 212-082-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only