missing translation for 'onlineSavingsMsg'
Learn More

DL-Mevalonolactone 98.0+%, TCI America™

Catalog No. D0587100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0587100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0587100MG Supplier TCI America Supplier No. D0587100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 674-26-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00006648
InChI Key JYVXNLLUYHCIIH-UHFFFAOYNA-N
Synonym mevalonolactone, dl-mevalonic acid lactone, dl-mevalonolactone, 4-hydroxy-4-methyltetrahydro-2h-pyran-2-one, mevalonic acid lactone, mevalonic lactone, dl-mevalolactone, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl-, .+/-., 1-tetrahydro-4-hydroxy-4-methyl-2h-pyran-2-one
PubChem CID 10428
IUPAC Name 4-hydroxy-4-methyloxan-2-one
SMILES CC1(O)CCOC(=O)C1

Specifications

Melting Point 28°C
Color White-Yellow
Boiling Point 150°C
Quantity 100 mg
Formula Weight 130.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material DL-Mevalonolactone
TSCA No
Recommended Storage Refrigerator