missing translation for 'onlineSavingsMsg'
Learn More

DL-Methionine 99.0+%, TCI America™

$46.20 - $300.33

Chemical Identifiers

CAS 59-51-8
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063096
InChI Key FFEARJCKVFRZRR-UHFFFAOYNA-N
Synonym dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione
PubChem CID 876
ChEBI CHEBI:16811
IUPAC Name 2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCCC(N)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M046325G
View Documents
TCI America
M046325G
25 g
Each for $46.20
Only null left
 
M0463500G
View Documents
TCI America
M0463500G
500 g
Each for $300.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

59-51-8
149.21
FFEARJCKVFRZRR-UHFFFAOYNA-N
876
2-amino-4-(methylsulfanyl)butanoic acid
C5H11NO2S
MFCD00063096
dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione
CHEBI:16811
CSCCC(N)C(O)=O

Specifications

59-51-8
5.6 to 6.1 (1% soln.H2O)
MFCD00063096
dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione
CSCCC(N)C(O)=O
149.21
CHEBI:16811
≥99.0% (HPLC,T)
DL-Methionine
White
C5H11NO2S
25 g
FFEARJCKVFRZRR-UHFFFAOYNA-N
2-amino-4-(methylsulfanyl)butanoic acid
876
149.21
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1254&(2)-1467

RTECSNumber : PD0456000

TSCA : Yes

SDS
Documents

Documents