missing translation for 'onlineSavingsMsg'
Learn More

DL-Methionine 99.0+%, TCI America™

Catalog No. M0463500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0463500G 500 g
M046325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0463500G Supplier TCI America Supplier No. M0463500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59-51-8
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063096
InChI Key FFEARJCKVFRZRR-UHFFFAOYNA-N
Synonym dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione
PubChem CID 876
ChEBI CHEBI:16811
IUPAC Name 2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCCC(N)C(O)=O

Specifications

Color White
pH 5.6 to 6.1 (1% soln.H2O)
Quantity 500 g
Formula Weight 149.21
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Methionine
EINECSNumber (2)-1254&(2)-1467
RTECSNumber PD0456000
TSCA Yes