missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Methionine 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| pH | 5.6 to 6.1 (1% soln.H2O) |
| Quantity | 25 g |
| Formula Weight | 149.21 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | DL-Methionine |
Safety and Handling
| EINECSNumber | (2)-1254&(2)-1467 |
| RTECSNumber | PD0456000 |
| TSCA | Yes |