missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Methionine Sulfone 99.0+%, TCI America™
Supplier: TCI America M01005G
Specifications
| DL-Methionine Sulfone | |
| 250°C | |
| C5H11NO4S | |
| 5 g | |
| UCUNFLYVYCGDHP-UHFFFAOYNA-N | |
| 2-amino-4-methanesulfonylbutanoic acid | |
| 69961 | |
| ≥99.0% (T) |
| 820-10-0 | |
| White | |
| MFCD00025079 | |
| dl-methionine sulfone, methionine sulfone, 2-amino-4-methylsulfonyl butanoic acid, butanoic acid, 2-amino-4-methylsulfonyl, d,l-methionine sulfone, methioninesulfone, dl-2-amino-4-methylsulfonyl butanoic acid, 2-amino-4-methylsulfonyl butyric acid, methionine, s,s-dioxide, butyric acid, 2-amino-4-methylsulfonyl-, dl | |
| CS(=O)(=O)CCC(N)C(O)=O | |
| 181.21 | |
| 181.21 | |
| Crystalline Powder |
Chemical Identifiers
| 820-10-0 | |
| 181.21 | |
| UCUNFLYVYCGDHP-UHFFFAOYNA-N | |
| 69961 | |
| CS(=O)(=O)CCC(N)C(O)=O |
| C5H11NO4S | |
| MFCD00025079 | |
| dl-methionine sulfone, methionine sulfone, 2-amino-4-methylsulfonyl butanoic acid, butanoic acid, 2-amino-4-methylsulfonyl, d,l-methionine sulfone, methioninesulfone, dl-2-amino-4-methylsulfonyl butanoic acid, 2-amino-4-methylsulfonyl butyric acid, methionine, s,s-dioxide, butyric acid, 2-amino-4-methylsulfonyl-, dl | |
| 2-amino-4-methanesulfonylbutanoic acid |
Safety and Handling
TSCA : Yes