missing translation for 'onlineSavingsMsg'
Learn More

DL-Methionine Sulfone 99.0+%, TCI America™

Catalog No. M01001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M01001G 1 g
M01005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M01001G Supplier TCI America Supplier No. M01001G
Only null left

Chemical Identifiers

CAS 820-10-0
Molecular Formula C5H11NO4S
Molecular Weight (g/mol) 181.21
MDL Number MFCD00025079
InChI Key UCUNFLYVYCGDHP-UHFFFAOYNA-N
Synonym dl-methionine sulfone, methionine sulfone, 2-amino-4-methylsulfonyl butanoic acid, butanoic acid, 2-amino-4-methylsulfonyl, d,l-methionine sulfone, methioninesulfone, dl-2-amino-4-methylsulfonyl butanoic acid, 2-amino-4-methylsulfonyl butyric acid, methionine, s,s-dioxide, butyric acid, 2-amino-4-methylsulfonyl-, dl
PubChem CID 69961
IUPAC Name 2-amino-4-methanesulfonylbutanoic acid
SMILES CS(=O)(=O)CCC(N)C(O)=O

Specifications

Melting Point 250°C
Color White
Quantity 1 g
Formula Weight 181.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Methionine Sulfone