missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Methionine sulfone, 98%
CAS: 820-10-0 | C5H11NO4S | 181.21 g/mol
Supplier: Thermo Scientific Chemicals B2509403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| DL-Methionine sulfone | |
| C5H11NO4S | |
| dl-methionine sulfone, methionine sulfone, 2-amino-4-methylsulfonyl butanoic acid, butanoic acid, 2-amino-4-methylsulfonyl, d,l-methionine sulfone, methioninesulfone, dl-2-amino-4-methylsulfonyl butanoic acid, 2-amino-4-methylsulfonyl butyric acid, methionine, s,s-dioxide, butyric acid, 2-amino-4-methylsulfonyl-, dl | |
| CS(=O)(=O)CCC(N)C(O)=O | |
| 1 g | |
| 69961 | |
| 98% |
| 820-10-0 | |
| MFCD00025079 | |
| UCUNFLYVYCGDHP-UHFFFAOYNA-N | |
| 2-amino-4-methylsulfonylbutanoic acid | |
| 181.21 | |
| 181.21 |
Chemical Identifiers
| 820-10-0 | |
| 181.21 | |
| UCUNFLYVYCGDHP-UHFFFAOYNA-N | |
| 69961 | |
| CS(=O)(=O)CCC(N)C(O)=O |
| C5H11NO4S | |
| MFCD00025079 | |
| dl-methionine sulfone, methionine sulfone, 2-amino-4-methylsulfonyl butanoic acid, butanoic acid, 2-amino-4-methylsulfonyl, d,l-methionine sulfone, methioninesulfone, dl-2-amino-4-methylsulfonyl butanoic acid, 2-amino-4-methylsulfonyl butyric acid, methionine, s,s-dioxide, butyric acid, 2-amino-4-methylsulfonyl-, dl | |
| 2-amino-4-methylsulfonylbutanoic acid |
Safety and Handling
EINECSNumber : 212-466-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only