missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Methionine, 99%
Description
Production of volatile compounds related to the flavour of foods from the Strecker degradation of DL-methionine. Addition of methionine in appropriate amounts to these foods might be expected to improve protein value. Economically, DL-methionine would be preferable.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Production of volatile compounds related to the flavour of foods from the Strecker degradation of DL-methionine. Addition of methionine in appropriate amounts to these foods might be expected to improve protein value. Economically, DL-methionine would be preferable.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Production of volatile compounds related to the flavour of foods from the Strecker degradation of DL-methionine. Addition of methionine in appropriate amounts to these foods might be expected to improve protein value. Economically, DL-methionine would be preferable.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼272°C (decomposition) |
| Beilstein | 636185 |
| Merck Index | 14,5975 |
| Solubility Information | Soluble in water. |
| Density | 1.34 |
| IUPAC Name | 2-amino-4-(methylsulfanyl)butanoic acid |
| Quantity | 1000 g |
| Formula Weight | 149.21 |
| Percent Purity | 99% |
| Chemical Name or Material | DL-Methionine |
Safety and Handling
| Hazard Category | H500 |
| EINECSNumber | 200-432-1 |
| RTECSNumber | PD0456000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only