missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Methionine, 99+%
CAS: 59-51-8 | C5H11NO2S | 149.21 g/mol
$99.90 - $176.96
Chemical Identifiers
| CAS | 59-51-8 |
|---|---|
| Molecular Formula | C5H11NO2S |
| Molecular Weight (g/mol) | 149.21 |
| MDL Number | MFCD00063096 |
| InChI Key | FFEARJCKVFRZRR-UHFFFAOYNA-N |
| Synonym | dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione |
| PubChem CID | 876 |
| ChEBI | CHEBI:16811 |
| SMILES | CSCCC(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC125651000
|
Thermo Scientific Chemicals
125651000 |
100 g | Plastic Bottle |
Each for $99.90
|
|
||||
|
AC125652500
|
Thermo Scientific Chemicals
125652500 |
250 g | Plastic Bottle |
Each for $120.86
|
|
||||
|
AC125650010
|
Thermo Scientific Chemicals
125650010 |
1 kg | Plastic Bottle |
Each for $176.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 59-51-8 | |
| 149.21 | |
| FFEARJCKVFRZRR-UHFFFAOYNA-N | |
| 876 | |
| CSCCC(N)C(O)=O |
| C5H11NO2S | |
| MFCD00063096 | |
| dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione | |
| CHEBI:16811 |
Specifications
| 1000ppm max. | |
| 5.4 to 6.1 (2% w/v aq. soln.) | |
| 200ppm max. | |
| Authentic | |
| 99+% | |
| C5H11NO2S | |
| CH3SCH2CH2CH(NH2)CO2H | |
| 04, II, 938 | |
| 95% min. (c=10, 2N HCl, 430nm) 1cm cell | |
| 0.1% max. | |
| FFEARJCKVFRZRR-UHFFFAOYNA-N | |
| CSCCC(N)C(O)=O | |
| 149.21 | |
| 876 | |
| 149.21 | |
| Crystalline Powder or Crystals |
| 59-51-8 | |
| White | |
| 10ppm max. | |
| 0.5% max. (1 g, 105°C) | |
| 0.2% max. (TLC) | |
| Plastic Bottle | |
| MFCD00063096 | |
| 200ppm max. | |
| 15, 6047 | |
| dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione | |
| 1.34g/mL | |
| 2-amino-4-methylsulfanylbutanoic acid | |
| 100 g | |
| CHEBI:16811 | |
| 99.0 to 101.0% (on dried substance) | |
| DL-Methionine, 99+% |
Safety and Handling
EINECSNumber : 200-432-1
RTECSNumber : PD0456000
TSCA : TSCA
RUO – Research Use Only