missing translation for 'onlineSavingsMsg'
Learn More

DL-Menthol, 98+%

CAS: 89-78-1 | C10H20O | 156.269 g/mol

Supplier:  Thermo Scientific Chemicals A1809822

Quantity 100 g

 View more versions of this product

Catalog No. AAA1809822


Only null left

Description

Description

Used in perfumery to give power, lift, and freshness. For use in toothpaste, cough drops, mouthwash, ice cream, chewing gum, tobacco, medical plaster, tonic, and shampoos. Because of its cooling action, it is to be found in a wide range of medicines treating sore throats and mouth irritations. It is also used in treatments for minor aches and sprains, and in nasal decongestants. It is found in some oral hygiene products to combat bad breath. Of course, it is present in a number of chewing gums. It was first added to cigarettes in the 1920s and 1930s (e.g. Spud and Kool brands) for both its flavor and also to reduce irritation, especially to the throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, paraffin oil and water, 434.5 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

89-78-1
156.269
NOOLISFMXDJSKH-IVZWLZJFSA-N
6566020
CC1CCC(C(C1)O)C(C)C
C10H20O
MFCD00001484
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol

Specifications

DL-Menthol
33°C to 36°C
212°C to 216°C
Sweet
1.4615
MFCD00001484
14,5837
Soluble in alcohol,paraffin oil and water,434.5mg/L @ 25°C (est).
CC1CCC(C(C1)O)C(C)C
156.269
156.27
89-78-1
0.89
93°C (199°F)
C10H20O
100 g
3194263
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
NOOLISFMXDJSKH-IVZWLZJFSA-N
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
6566020
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-939-0

RTECSNumber : OT0350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions