missing translation for 'onlineSavingsMsg'
Learn More

DL-Menthol, 98+%

Catalog No. AAA1809818
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1809818 50 g
AAA1809822 100 g
AAA1809830 250 g
AAA180980B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1809818 Supplier Thermo Scientific Chemicals Supplier No. A1809818

CAS: 89-78-1 | C10H20O | 156.269 g/mol

Used in perfumery to give power, lift, and freshness. For use in toothpaste, cough drops, mouthwash, ice cream, chewing gum, tobacco, medical plaster, tonic, and shampoos. Because of its cooling action, it is to be found in a wide range of medicines treating sore throats and mouth irritations. It is also used in treatments for minor aches and sprains, and in nasal decongestants. It is found in some oral hygiene products to combat bad breath. Of course, it is present in a number of chewing gums. It was first added to cigarettes in the 1920s and 1930s (e.g. Spud and Kool brands) for both its flavor and also to reduce irritation, especially to the throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in perfumery to give power, lift, and freshness. For use in toothpaste, cough drops, mouthwash, ice cream, chewing gum, tobacco, medical plaster, tonic, and shampoos. Because of its ″cooling″ action, it is to be found in a wide range of medicines treating sore throats and mouth irritations. It is also used in treatments for minor aches and sprains, and in nasal decongestants. It is found in some oral hygiene products to combat ″bad breath″. Of course, it is present in a number of chewing gums. It was first added to cigarettes in the 1920s and 1930s (e.g. ″Spud″ and ″Kool″ brands) for both its flavor and also to reduce irritation, especially to the throat.

Solubility
Soluble in alcohol, paraffin oil and water, 434.5 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00001484
InChI Key NOOLISFMXDJSKH-IVZWLZJFSA-N
Synonym menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
PubChem CID 6566020
IUPAC Name (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Melting Point 33°C to 36°C
Density 0.89
Boiling Point 212°C to 216°C
Flash Point 93°C (199°F)
Odor Sweet
Refractive Index 1.4615
Quantity 50 g
Beilstein 3194263
Merck Index 14,5837
Solubility Information Soluble in alcohol,paraffin oil and water,434.5mg/L @ 25°C (est).
Formula Weight 156.27
Percent Purity ≥98%
Chemical Name or Material DL-Menthol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-939-0
RTECSNumber OT0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only