missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Mandelic Acid 99.0+%, TCI America™
Supplier: TCI America M0038500G
Specifications
| DL-Mandelic Acid | |
| 90-64-2 | |
| C8H8O3 | |
| mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid | |
| C1=CC=C(C=C1)C(C(=O)O)O | |
| 152.149 | |
| CHEBI:35825 | |
| ≥99.0% (GC,T) | |
| Crystalline Powder |
| 121°C | |
| White-Yellow | |
| MFCD00064250 | |
| IWYDHOAUDWTVEP-UHFFFAOYSA-N | |
| 2-hydroxy-2-phenylacetic acid | |
| 1292 | |
| 152.15 | |
| 500 g |
Chemical Identifiers
| 90-64-2 | |
| 152.149 | |
| IWYDHOAUDWTVEP-UHFFFAOYSA-N | |
| 1292 | |
| 2-hydroxy-2-phenylacetic acid |
| C8H8O3 | |
| MFCD00064250 | |
| mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid | |
| CHEBI:35825 | |
| C1=CC=C(C=C1)C(C(=O)O)O |
Safety and Handling
EINECSNumber : (3)-1764
RTECSNumber : OO6300000
TSCA : Yes