missing translation for 'onlineSavingsMsg'
Learn More

DL-Mandelic Acid 99.0+%, TCI America™

Catalog No. M003825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M003825G 25 g
M0038500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M003825G Supplier TCI America Supplier No. M003825G

Chemical Identifiers

CAS 90-64-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00064250
InChI Key IWYDHOAUDWTVEP-UHFFFAOYSA-N
Synonym mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid
PubChem CID 1292
ChEBI CHEBI:35825
IUPAC Name 2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

Melting Point 121°C
Color White-Yellow
Formula Weight 152.15
Percent Purity ≥99.0% (GC,T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material DL-Mandelic Acid
EINECSNumber (3)-1764
RTECSNumber OO6300000
TSCA Yes