missing translation for 'onlineSavingsMsg'
Learn More

DL-Mandelic Acid, 98%, Spectrum™ Chemical

Catalog No. p-7165919
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
12 kg
2.5 kg
50 kg
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18610299 100 g Amber Glass Bottle
18610301 500 g Amber Glass Bottle
18610298 2.5 kg Poly Pail
18610300 12 kg Poly Pail
18610302 50 kg Fiber Drum
5 options

Catalog No. 18610299

Supplier: Spectrum Chemical Mfg Cor MA160100GM

Only null left

Chemical Identifiers

CAS 611-72-3

Specifications

CAS Max % 1
Percent Purity 98%
Grade Reagent
Quantity 100 g
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.