missing translation for 'onlineSavingsMsg'
Learn More

DL-Mandelic Acid, 98%, Spectrum™ Chemical

Catalog No. 18610302
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610302 50 kg Fiber Drum
18610299 100 g Amber Glass Bottle
18610301 500 g Amber Glass Bottle
18610298 2.5 kg Poly Pail
18610300 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610302 Supplier Spectrum Chemical Mfg Cor Supplier No. MA16050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 611-72-3

Specifications

CAS Max % 1
Quantity 50 kg
Packaging Fiber Drum
Percent Purity 98%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.