missing translation for 'onlineSavingsMsg'
Learn More

DL-Mandelic Acid, 98%, Spectrum™ Chemical

Catalog No. 18610299
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610299 100 g Amber Glass Bottle
18610301 500 g Amber Glass Bottle
18610298 2.5 kg Poly Pail
18610300 12 kg Poly Pail
18610302 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610299 Supplier Spectrum Chemical Mfg Cor Supplier No. MA160100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 611-72-3

Specifications

CAS Max % 1
Percent Purity 98%
Grade Reagent
Quantity 100 g
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.