missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich DL-Malic acid
Description
DL-Malic acid
Spécifications
Spécifications
| Melting Point | 131°C to 133°C (lit.) |
| CAS | 6915-15-7 |
| Molecular Formula | C4H6O5 |
| Linear Formula | HO2CCH2CH(OH)CO2H |
| MDL Number | MFCD00064212 |
| Synonym | (+/-)-2-Hydroxysuccinic acid; DL-Hydroxybutanedioic acid |
| Molecular Weight (g/mol) | 134.09 |
| Percent Purity | ≥98% (capillary GC) |
| Quantity | 1 kg |