missing translation for 'onlineSavingsMsg'
Learn More

DL-m-Tyrosine 98.0+%, TCI America™

Catalog No. T07871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07871G 1 g
T07875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07871G Supplier TCI America Supplier No. T07871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 775-06-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002597
InChI Key JZKXXXDKRQWDET-UHFFFAOYNA-N
Synonym dl-m-tyrosine, 2-amino-3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl-dl-alanine, m-tyrosine, dl, d,l-metatyrosine, 3-hydroxy-dl-phenylalanine, dl-meta-tyrosine, dl-phenylalanine, 3-hydroxy, 3-m-hydroxyphenyl-dl-alanine, dl-3-tyrosine
PubChem CID 13052
IUPAC Name 2-amino-3-(3-hydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=CC(O)=C1)C(O)=O

Specifications

Melting Point 285°C
Quantity 1 g
Formula Weight 181.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-m-Tyrosine
RTECSNumber YP2278000
TSCA No