missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine monohydrochloride, 99%

Catalog No. AC413370250
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413370250 25 g Glass Bottle
AC413371000 100 g Glass Bottle
AC413370010 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC413370250 Supplier Thermo Scientific Chemicals Supplier No. 413370250
Only null left

CAS: 70-53-1 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70-53-1
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064563
InChI Key BVHLGVCQOALMSV-UHFFFAOYSA-N
Synonym dl-lysine monohydrochloride, 2,6-diaminohexanoic acid hydrochloride, dl-lysine hydrochloride, lysine monohydrochloride, dl-lysine mono hydrochloride, lysine, hydrochloride 1:1, lysine, monohydrochloride, dl-2,6-diaminohexanoic acid monohydrochloride, lysine, dl, dl-lysinemonohydrochloride
PubChem CID 12047
IUPAC Name 2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCCC(N)C(O)=O

Specifications

Melting Point 267.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula NH2(CH2)4CH(NH2)COO·HCl
Specific Rotation −1° to +1° (20°C, 589nm) (c=1, water)
Merck Index 14, 5636
Quantity 25 g
Formula Weight 182.65
Percent Purity 99%
Physical Form Powder, Granules or Chunks
Chemical Name or Material DL-Lysine monohydrochloride, 99%
Show More Show Less
EINECSNumber 200-739-0
TSCA TSCA

RUO – Research Use Only