missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Lysine monohydrate, 98+%
CAS: 885701-25-7 | C6H16N2O3 | 164.21 g/mol
$65.32 - $192.42
Chemical Identifiers
| CAS | 885701-25-7 |
|---|---|
| Molecular Formula | C6H16N2O3 |
| Molecular Weight (g/mol) | 164.21 |
| MDL Number | MFCD00151036 |
| InChI Key | HZRUTVAFDWTKGD-UHFFFAOYNA-N |
| Synonym | dl-lysine monohydrate, 2,6-diaminohexanoic acid hydrate, lysine hydrate, l-2,6-diaminohexanoic acid, dl-lysine hydrate, 2,6-diaminohexanoic acid,hydrate |
| PubChem CID | 2734605 |
| IUPAC Name | 2,6-diaminohexanoic acid;hydrate |
| SMILES | O.NCCCCC(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0757606
|
Thermo Scientific Chemicals
L0757606 |
5 g |
Each for $65.32
|
|
|||||
|
AAL0757614
|
Thermo Scientific Chemicals
L0757614 |
25 g |
Each for $192.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 885701-25-7 | |
| 164.21 | |
| HZRUTVAFDWTKGD-UHFFFAOYNA-N | |
| 2734605 | |
| O.NCCCCC(N)C(O)=O |
| C6H16N2O3 | |
| MFCD00151036 | |
| dl-lysine monohydrate, 2,6-diaminohexanoic acid hydrate, lysine hydrate, l-2,6-diaminohexanoic acid, dl-lysine hydrate, 2,6-diaminohexanoic acid,hydrate | |
| 2,6-diaminohexanoic acid;hydrate |
Specifications
| ∼110°C | |
| C6H16N2O3 | |
| 1616991 | |
| HZRUTVAFDWTKGD-UHFFFAOYNA-N | |
| 2,6-diaminohexanoic acid;hydrate | |
| 164.21 | |
| 164.21 (146.19 Anhydrous) | |
| DL-Lysine monohydrate |
| 885701-25-7 | |
| MFCD00151036 | |
| dl-lysine monohydrate, 2,6-diaminohexanoic acid hydrate, lysine hydrate, l-2,6-diaminohexanoic acid, dl-lysine hydrate, 2,6-diaminohexanoic acid,hydrate | |
| O.NCCCCC(N)C(O)=O | |
| 5 g | |
| 2734605 | |
| ≥98% |
Safety and Handling
EINECSNumber : 200-740-6
RTECSNumber : OL5540000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only