missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine monohydrate, 98+%

Catalog No. AAL0757614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0757614 25 g
AAL0757606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0757614 Supplier Thermo Scientific Chemicals Supplier No. L0757614
Only null left

CAS: 885701-25-7 | C6H16N2O3 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 885701-25-7
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151036
InChI Key HZRUTVAFDWTKGD-UHFFFAOYNA-N
Synonym dl-lysine monohydrate, 2,6-diaminohexanoic acid hydrate, lysine hydrate, l-2,6-diaminohexanoic acid, dl-lysine hydrate, 2,6-diaminohexanoic acid,hydrate
PubChem CID 2734605
IUPAC Name 2,6-diaminohexanoic acid;hydrate
SMILES O.NCCCCC(N)C(O)=O

Specifications

Melting Point ∼110°C
Beilstein 1616991
Quantity 25 g
Formula Weight 164.21 (146.19 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material DL-Lysine monohydrate
EINECSNumber 200-740-6
RTECSNumber OL5540000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only