missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine monohydrate, 97%

Catalog No. AC365650250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC365650250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC365650250 Supplier Thermo Scientific Chemicals Supplier No. 365650250
Only null left

CAS: 885701-25-7 | C6H16N2O3 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 885701-25-7
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151036
InChI Key HZRUTVAFDWTKGD-UHFFFAOYNA-N
Synonym dl-lysine monohydrate, 2,6-diaminohexanoic acid hydrate, lysine hydrate, l-2,6-diaminohexanoic acid, dl-lysine hydrate, 2,6-diaminohexanoic acid,hydrate
PubChem CID 2734605
IUPAC Name 2,6-diaminohexanoic acid;hydrate
SMILES O.NCCCCC(N)C(O)=O

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Loss on Drying 7% to 12% (105°C, 2 hrs)
Packaging Glass Bottle
Linear Formula HN(CH)4CH(NH)COH
Quantity 25 g
Formula Weight 164.21
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material DL-Lysine monohydrate, 98%
Show More Show Less

RUO – Research Use Only