missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Leucine, 99+%
CAS: 328-39-2 | C6H13NO2 | 131.18 g/mol
$549.11 - $549.11
Chemical Identifiers
| CAS | 328-39-2 |
|---|---|
| Molecular Formula | C6H13NO2 |
| Molecular Weight (g/mol) | 131.18 |
| MDL Number | MFCD00063087 |
| InChI Key | ROHFNLRQFUQHCH-UHFFFAOYNA-N |
| Synonym | dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d |
| PubChem CID | 857 |
| ChEBI | CHEBI:25017 |
| IUPAC Name | 2-amino-4-methylpentanoic acid |
| SMILES | CC(C)CC(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC172132500
|
Thermo Scientific Chemicals
172132500 |
250 g | Plastic Bottle |
Each for $549.11
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 328-39-2 | |
| 131.18 | |
| ROHFNLRQFUQHCH-UHFFFAOYNA-N | |
| 857 | |
| 2-amino-4-methylpentanoic acid |
| C6H13NO2 | |
| MFCD00063087 | |
| dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d | |
| CHEBI:25017 | |
| CC(C)CC(N)C(O)=O |
Specifications
| 328-39-2 | |
| 20ppm max. | |
| 0.3% max. | |
| C6H13NO2 | |
| (CH3)2CHCH2CH(NH2)CO2H | |
| 04, 447 | |
| 0.1% max. | |
| ROHFNLRQFUQHCH-UHFFFAOYNA-N | |
| 2-amino-4-methylpentanoic acid | |
| 250 g | |
| CHEBI:25017 | |
| 99.0 to 101.0% (on dry substance) | |
| DL-Leucine, 99+% |
| White | |
| Authentic | |
| 99+% | |
| Plastic Bottle | |
| MFCD00063087 | |
| 14, 5451 | |
| dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d | |
| CC(C)CC(N)C(O)=O | |
| 131.18 | |
| 857 | |
| 131.17 | |
| Fine Crystalline Powder |
Safety and Handling
EINECSNumber : 206-328-2
TSCA : TSCA
RUO – Research Use Only