missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Leucine, 99+%

Catalog No. AC172130250
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172130250 25 g Plastic Bottle
AC172132500 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172130250 Supplier Thermo Scientific Chemicals Supplier No. 172130250
Only null left

CAS: 328-39-2 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 328-39-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063087
InChI Key ROHFNLRQFUQHCH-UHFFFAOYNA-N
Synonym dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,d
PubChem CID 857
ChEBI CHEBI:25017
IUPAC Name 2-amino-4-methylpentanoic acid
SMILES CC(C)CC(N)C(O)=O

Specifications

Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Loss on Drying 0.3% max.
Assay Percent Range 99+%
Packaging Plastic Bottle
Linear Formula (CH3)2CHCH2CH(NH2)CO2H
Beilstein 04, 447
Merck Index 14, 5451
Sulfated Ash 0.1% max.
Quantity 25 g
Formula Weight 131.17
Percent Purity 99.0 to 101.0% (on dry substance)
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Leucine, 99+%
Show More Show Less
EINECSNumber 206-328-2
TSCA TSCA

RUO – Research Use Only