missing translation for 'onlineSavingsMsg'
Learn More

DL-Leucic Acid 98.0+%, TCI America™

Catalog No. p-7131822
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L010425G 25 g
L0104500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L010425G Supplier TCI America Supplier No. L010425G
Only null left

Chemical Identifiers

CAS 498-36-2
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00004246
InChI Key LVRFTAZAXQPQHI-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methylvaleric acid, 2-hydroxyisocaproic acid, leucic acid, dl-leucic acid, alpha-hydroxyisocaproic acid, leucinic acid, 2-hydroxy-4-methyl-pentanoic acid, pentanoic acid, 2-hydroxy-4-methyl, dl-2-hydroxyisocaproic acid, leucate
PubChem CID 92779
ChEBI CHEBI:59783
IUPAC Name 2-hydroxy-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)O

Specifications

Melting Point 75°C
Color White
Quantity 25 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Leucic Acid
EINECSNumber (2)-1340
TSCA Yes