missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactide, 99%

CAS: 95-96-5 | C6H8O4 | 144.13 g/mol

$427.24 - $427.24

Chemical Identifiers

CAS 95-96-5
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00011685,MFCD00082566
InChI Key JJTUDXZGHPGLLC-UHFFFAOYNA-N
Synonym dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
PubChem CID 7272
IUPAC Name 3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC1OC(=O)C(C)OC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC376851000
SDS View Documents
Thermo Scientific Chemicals
376851000
100 g Glass bottle
Each for $427.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-96-5
144.13
JJTUDXZGHPGLLC-UHFFFAOYNA-N
7272
CC1OC(=O)C(C)OC1=O
C6H8O4
MFCD00011685,MFCD00082566
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
3,6-dimethyl-1,4-dioxane-2,5-dione

Specifications

122°C to 126°C
White
100.0
100 g
Glass bottle
98.5% min. (GC)
MFCD00011685,MFCD00082566
JJTUDXZGHPGLLC-UHFFFAOYNA-N
3,6-dimethyl-1,4-dioxane-2,5-dione
7272
99%
95-96-5
98.5
138°C to 142°C (8.0 mmHg)
Authentic
Crystalline Powder
C6H8O4
dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
CC1OC(=O)C(C)OC1=O
144.13
144.13
DL-Lactide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

EINECSNumber : 202-468-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only