missing translation for 'onlineSavingsMsg'
Learn More

DL-Lactide, 99%

Catalog No. AC376850250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC376850250 25 g Glass bottle
AC376851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC376850250 Supplier Thermo Scientific Chemicals Supplier No. 376850250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-96-5 | C6H8O4 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-96-5
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00011685,MFCD00082566
InChI Key JJTUDXZGHPGLLC-UHFFFAOYNA-N
Synonym dl-lactide, lactide, dilactide, 1,4-dioxane-2,5-dione, 3,6-dimethyl, 3,6-dimethyl-2,5-dioxo-1,4-dioxane, lactic acid, bimol. cyclic ester, propanoic acid, 2-hydroxy-, bimol. cyclic ester, p-dioxane-2,5-dione, 3,6-dimethyl, d +-lactide, --l-dilactide
PubChem CID 7272
IUPAC Name 3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC1OC(=O)C(C)OC1=O

Specifications

Melting Point 122°C to 126°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Quantity 25 g
Boiling Point 138°C to 142°C (8.0mmHg), 138°C to 142°C (8.0 mmHg)
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98.5% min. (GC)
Formula Weight 144.13
Percent Purity 99%
Chemical Name or Material DL-Lactide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 202-468-3

RUO – Research Use Only